Assay Details
Gene Symbol: MAPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099826.4 | XP_001099826.2 | 1795 | 80 | ||
XM_015118782.2 | XP_014974268.2 | 2172 | 80 | |||
XM_015118783.2 | XP_014974269.2 | 2154 | 80 | |||
XM_015118784.2 | XP_014974270.2 | 1837 | 80 | |||
XM_028836095.1 | XP_028691928.1 | 1937 | 80 | |||
XM_028836096.1 | XP_028691929.1 | 1788 | 80 | |||
XM_028836097.1 | XP_028691930.1 | 1567 | 80 | |||
XM_028836098.1 | XP_028691931.1 | 1332 | 80 | |||
GenBank mRNA | JV656104.1 | - | 1603 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
