Assay Details
Gene Symbol: LOC714737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148079.2 | XP_015003565.2 | 116 | 144 | ||
XM_028842703.1 | XP_028698536.1 | 151 | 144 | |||
XM_028842704.1 | XP_028698537.1 | 127 | 144 | |||
GenBank mRNA | JU335500.1 | - | 136 | 144 | ||
JU335501.1 | - | 136 | 144 | |||
JV600905.1 | - | 441 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015148079.2, XM_028842703.1, XM_028842704.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
