Assay Details
Gene Symbol: LOC714737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148079.2 | XP_015003565.2 | 1328 | 126 | ||
XM_028842703.1 | XP_028698536.1 | 1363 | 126 | |||
XM_028842704.1 | XP_028698537.1 | 1339 | 126 | |||
GenBank mRNA | JU335500.1 | - | 1348 | 126 | ||
JU335501.1 | - | 1348 | 126 | |||
JV600905.1 | - | 1653 | 126 |
Gene Symbol: PSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028826803.1 | XP_028682636.1 | 1152 | 126 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
