Assay Details
| Gene Symbol: RPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195652.1 | NP_001182581.1 | 336 | 76 | ||
| GenBank mRNA | CO581132.1 | - | 340 | 76 | ||
| JU326689.1 | - | 298 | 76 | |||
| JU476699.1 | - | 388 | 76 | |||
| JV044390.1 | - | 391 | 76 | |||
| JV590057.1 | - | 358 | 76 | |||
| JV658162.1 | - | 389 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPN1
Genomic Map