Assay Details
| Gene Symbol: MAPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112396.4 | XP_001112396.3 | 623 | 63 | ||
| XM_001112462.4 | XP_001112462.2 | 623 | 63 | |||
| XM_028827557.1 | XP_028683390.1 | 493 | 63 | |||
| GenBank mRNA | JU326663.1 | - | 248 | 63 | ||
| JV601778.1 | - | 458 | 63 | |||
More Information
Set Membership: |
|

MAPK12
Genomic Map