Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110268.4 | XP_001110268.1 | 2276 | 72 | ||
| GenBank mRNA | KT382282.1 | - | 564 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC10A1| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110268.4 | XP_001110268.1 | 2276 | 72 | ||
| GenBank mRNA | KT382282.1 | - | 564 | 72 | ||
Set Membership: |
|