Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103789.4 | XP_001103789.2 | 1726 | 73 | ||
| XM_002804509.3 | XP_002804555.2 | 1798 | 73 | |||
More Information
Set Membership: |
|

SLC22A5
Genomic Map