Assay Details
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266152.1 | NP_001253081.1 | 135 | 89 | ||
XM_015135063.2 | XP_014990549.1 | 287 | 89 | |||
XM_015135064.2 | XP_014990550.1 | 356 | 89 | |||
XM_015135066.2 | XP_014990552.1 | 198 | 89 | |||
XM_015135068.2 | XP_014990554.1 | 287 | 89 | |||
XM_015135069.2 | XP_014990555.1 | 373 | 89 | |||
GenBank mRNA | JU322533.1 | - | 133 | 89 | ||
JU470814.1 | - | 135 | 89 | |||
JV045286.1 | - | 135 | 89 | |||
JV639470.1 | - | 138 | 89 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
