Assay Details
Gene Symbol: LOC695267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146698.2 | XP_015002184.2 | 6491 | 90 | ||
XM_015146699.2 | XP_015002185.2 | 6095 | 90 | |||
XM_015146702.2 | XP_015002188.2 | 6033 | 90 | |||
XM_015146704.2 | XP_015002190.2 | 6062 | 90 | |||
XM_015146705.2 | XP_015002191.2 | 6413 | 90 | |||
XM_015146706.2 | XP_015002192.2 | 6395 | 90 | |||
XM_015146707.2 | XP_015002193.2 | 6365 | 90 | |||
XM_015146708.2 | XP_015002194.2 | 6320 | 90 | |||
XM_015146709.2 | XP_015002195.2 | 6269 | 90 | |||
XM_015146710.2 | XP_015002196.2 | 5567 | 90 | |||
XM_015146711.2 | XP_015002197.2 | 5396 | 90 | |||
XM_015146713.2 | XP_015002199.2 | 5315 | 90 | |||
XM_015146714.2 | XP_015002200.2 | 5219 | 90 | |||
XM_015146715.2 | XP_015002201.2 | 5093 | 90 | |||
XM_028826127.1 | XP_028681960.1 | 6228 | 90 | |||
XM_028826128.1 | XP_028681961.1 | 5441 | 90 | |||
XM_028826130.1 | XP_028681963.1 | 5189 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
