Assay Details
| Gene Symbol: TFAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097656.4 | XP_001097656.1 | 1055 | 127 | ||
| XM_015147590.2 | XP_015003076.1 | 994 | 127 | |||
| GenBank mRNA | JV480081.1 | - | 492 | 127 | ||
| JV703614.1 | - | 653 | 127 | |||
More Information
Set Membership: |
|

TFAM
Genomic Map