Assay Details
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083645.4 | XP_001083645.1 | 651 | 162 | ||
XM_015145563.2 | XP_015001049.1 | 647 | 162 | |||
GenBank mRNA | JV592099.1 | - | 649 | 162 | ||
JV694734.1 | - | 648 | 162 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
