Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036685.1 | NP_001031762.1 | 2481 | 133 | ||
XM_015139554.2 | XP_014995040.2 | 2885 | 133 | |||
XM_028847940.1 | XP_028703773.1 | 3108 | 133 | |||
GenBank mRNA | AB445637.1 | - | 2481 | 133 | ||
AY525613.1 | - | 216 | 133 | |||
AY864735.1 | - | 2481 | 133 | |||
EU204934.1 | - | 2487 | 133 | |||
JV722246.1 | - | 168 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
