Assay Details
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266571.2 | NP_001253500.1 | 170 | 65 | ||
XM_015146232.2 | XP_015001718.2 | 195 | 65 | |||
XM_015146233.2 | XP_015001719.2 | 644 | 65 | |||
XM_015146235.2 | XP_015001721.2 | 302 | 65 | |||
XM_015146236.2 | XP_015001722.2 | 323 | 65 | |||
XM_015146238.2 | XP_015001724.2 | 550 | 65 | |||
GenBank mRNA | JU323957.1 | - | 236 | 65 | ||
JU472655.1 | - | 231 | 65 | |||
JV045382.1 | - | 170 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
