Assay Details
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099833.3 | XP_001099833.2 | 1864 | 80 | ||
| XM_015124992.2 | XP_014980478.2 | 2034 | 80 | |||
| XM_015124993.2 | XP_014980479.2 | 1866 | 80 | |||
| XM_028838820.1 | XP_028694653.1 | 3049 | 80 | |||
| XM_028838821.1 | XP_028694654.1 | 1696 | 80 | |||
| GenBank mRNA | JV660774.1 | - | 1028 | 80 | ||
More Information
Set Membership: |
|

MZF1
Genomic Map