Assay Details
Gene Symbol: LOC699378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU326120.1 | - | 72 | 132 |
Gene Symbol: VAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838178.1 | XP_028694011.1 | 1303 | 132 | ||
XM_028838179.1 | XP_028694012.1 | 1154 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
