Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204799.2 | NP_001191728.2 | 321 | 92 | ||
| GenBank mRNA | JU323979.1 | - | 281 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FPGT| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204799.2 | NP_001191728.2 | 321 | 92 | ||
| GenBank mRNA | JU323979.1 | - | 281 | 92 | ||
Set Membership: |
|