Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194043.1 | NP_001180972.1 | 539 | 83 | ||
| XM_015122285.2 | XP_014977771.1 | 470 | 83 | |||
| GenBank mRNA | JU327295.1 | - | 330 | 83 | ||
| JU473478.1 | - | 419 | 83 | |||
| JV635422.1 | - | 437 | 83 | |||
| JV636726.1 | - | 437 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MBD3
Genomic Map