Assay Details
| Gene Symbol: CSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257285.1 | NP_001244214.1 | 464 | 84 | ||
| XM_015118823.2 | XP_014974309.2 | 938 | 84 | |||
| XM_015118831.2 | XP_014974317.1 | 529 | 84 | |||
| XM_028843664.1 | XP_028699497.1 | 589 | 84 | |||
| GenBank mRNA | BQ807632.1 | - | 464 | 84 | ||
| BQ807767.1 | - | 288 | 84 | |||
| JU474399.1 | - | 494 | 84 | |||
| JV673835.1 | - | 484 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CSRP1
Genomic Map