Assay Details
| Gene Symbol: FGF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108934.4 | XP_001108934.1 | 2050 | 85 | ||
| GenBank mRNA | JU331316.1 | - | 164 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF11| Gene Symbol: FGF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108934.4 | XP_001108934.1 | 2050 | 85 | ||
| GenBank mRNA | JU331316.1 | - | 164 | 85 | ||
Set Membership: |
|