Assay Details
Gene Symbol: LOC719690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU334622.1 | - | 1897 | 116 | ||
JV047134.1 | - | 1924 | 116 | |||
JV652224.1 | - | 1944 | 116 |
Gene Symbol: MYO9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839737.1 | XP_028695570.1 | 1806 | 116 |

More Information
Set Membership: |
![]() ![]() |
