Assay Details
| Gene Symbol: NDUFB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194515.1 | NP_001181444.1 | 348 | 80 | ||
| GenBank mRNA | JU323496.1 | - | 292 | 80 | ||
| JV045227.1 | - | 287 | 80 | |||
| JV529407.1 | - | 349 | 80 | |||
| JV668363.1 | - | 334 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFB7
Genomic Map