Assay Details
| Gene Symbol: RAB27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083017.4 | XP_001083017.1 | 77 | |||
| XM_001083132.4 | XP_001083132.1 | 77 | ||||
| GenBank mRNA | JU326608.1 | - | 130 | 77 | ||
| JV589553.1 | - | 501 | 77 | |||
More Information
Set Membership: |
|

RAB27B
Genomic Map