Assay Details
Gene Symbol: SLC7A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266035.2 | NP_001252964.1 | 1028 | 82 | ||
XM_028827174.1 | XP_028683007.1 | 1060 | 82 | |||
GenBank mRNA | JU335999.1 | - | 1043 | 82 | ||
JV047983.1 | - | 1029 | 82 | |||
JV555356.1 | - | 1039 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
