Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265956.1 | NP_001252885.1 | 253 | 76 | ||
GenBank mRNA | JU335597.1 | - | 269 | 76 | ||
JU476509.1 | - | 253 | 76 | |||
JV047738.1 | - | 247 | 76 | |||
JV623044.1 | - | 272 | 76 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
