Assay Details
| Gene Symbol: CPT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289956.1 | NP_001276885.1 | 2113 | 103 | ||
| XM_015150649.2 | XP_015006135.2 | 2426 | 103 | |||
| XM_015150650.2 | XP_015006136.2 | 2127 | 103 | |||
| GenBank mRNA | JU326494.1 | - | 1461 | 103 | ||
| JU326495.1 | - | 1461 | 103 | |||
| JV044678.1 | - | 2047 | 103 | |||
| JV635559.1 | - | 2113 | 103 | |||
More Information
Set Membership: |
|

CPT1B
Genomic Map