Assay Details
Gene Symbol: DUSP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086701.4 | XP_001086701.4 | 795 | 87 | ||
GenBank mRNA | JV046358.1 | - | 557 | 87 | ||
JV636003.1 | - | 584 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
