Assay Details
| Gene Symbol: RPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258154.1 | NP_001245083.1 | 516 | 126 | ||
| XM_015137252.2 | XP_014992738.1 | 599 | 126 | |||
| GenBank mRNA | JU471973.1 | - | 516 | 126 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPF2
Genomic Map