Assay Details
Gene Symbol: POLDIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190901.2 | NP_001177830.2 | 817 | 86 | ||
XM_015150410.2 | XP_015005896.2 | 837 | 86 | |||
XM_015150411.2 | XP_015005897.2 | 871 | 86 | |||
GenBank mRNA | JU321178.1 | - | 881 | 86 | ||
JU471784.1 | - | 910 | 86 | |||
JU471785.1 | - | 823 | 86 | |||
JV044788.1 | - | 914 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
