Assay Details
| Gene Symbol: LZTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109770.4 | XP_001109770.1 | 599 | 98 | ||
| XM_001109817.2 | XP_001109817.1 | 619 | 98 | |||
| XM_028826780.1 | XP_028682613.1 | 2852 | 98 | |||
| XM_028826781.1 | XP_028682614.1 | 851 | 98 | |||
| XM_028826782.1 | XP_028682615.1 | 305 | 98 | |||
| XM_028826784.1 | XP_028682617.1 | 389 | 98 | |||
| GenBank mRNA | JU336724.1 | - | 469 | 98 | ||
| JU472734.1 | - | 453 | 98 | |||
| JV043749.1 | - | 451 | 98 | |||
| JV668537.1 | - | 479 | 98 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826782.1, XM_028826784.1 transcripts.
Set Membership: |
|

LZTS2
Genomic Map