Assay Details
Gene Symbol: CREB3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122496.2 | XP_014977982.2 | 1033 | 71 | ||
XM_028839065.1 | XP_028694898.1 | 1010 | 71 | |||
XM_028839066.1 | XP_028694899.1 | 926 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
