Assay Details
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139247.2 | XP_014994733.1 | 822 | 88 | ||
XM_015139250.2 | XP_014994736.1 | 823 | 88 | |||
XM_015139254.2 | XP_014994740.1 | 826 | 88 | |||
XM_015139261.2 | XP_014994747.1 | 783 | 88 | |||
XM_015139275.2 | XP_014994761.1 | 787 | 88 | |||
XM_028844223.1 | XP_028700056.1 | 780 | 88 | |||
XM_028844227.1 | XP_028700060.1 | 744 | 88 | |||
XM_028844233.1 | XP_028700066.1 | 782 | 88 | |||
XM_028844260.1 | XP_028700093.1 | 619 | 88 | |||
GenBank mRNA | JV725447.1 | - | 716 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
