Assay Details
Gene Symbol: UPK3BL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108997.4 | XP_001108997.2 | 1031 | 145 |

More Information
Set Membership: |
![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UPK3BL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108997.4 | XP_001108997.2 | 1031 | 145 |
Set Membership: |
![]() ![]() |