Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086489.4 | XP_001086489.3 | 800 | 124 | ||
| XM_028837801.1 | XP_028693634.1 | 800 | 124 | |||
| XM_028837802.1 | XP_028693635.1 | 369 | 124 | |||
| GenBank mRNA | JU321557.1 | - | 338 | 124 | ||
| JU470859.1 | - | 391 | 124 | |||
| JV045163.1 | - | 363 | 124 | |||
| JV529684.1 | - | 394 | 124 | |||
More Information
Set Membership: |
|

SEC11C
Genomic Map