Assay Details
Gene Symbol: CENPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261531.1 | NP_001248460.1 | 198 | 120 | ||
XM_015123292.2 | XP_014978778.1 | 120 | ||||
GenBank mRNA | JU477793.1 | - | 198 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261531.1, XM_015123292.2 transcripts.
Set Membership: |
![]() ![]() |
