Assay Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261531.1 | NP_001248460.1 | 1165 | 125 | ||
| XM_015123292.2 | XP_014978778.1 | 1182 | 125 | |||
| GenBank mRNA | JU477793.1 | - | 1165 | 125 | ||
More Information
Set Membership: |
|

CENPL
Genomic Map