Assay Details
Gene Symbol: DYNC2I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116677.2 | XP_014972163.1 | 933 | 165 | ||
XM_015116678.2 | XP_014972164.1 | 838 | 165 | |||
XM_028834760.1 | XP_028690593.1 | 1016 | 165 | |||
GenBank mRNA | JV632640.1 | - | 828 | 165 | ||
JV664827.1 | - | 816 | 165 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
