Assay Details
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082263.4 | XP_001082263.2 | 1359 | 100 | ||
XM_015135075.2 | XP_014990561.2 | 1359 | 100 | |||
XM_015135076.2 | XP_014990562.2 | 1359 | 100 | |||
GenBank mRNA | JU323286.1 | - | 1016 | 100 | ||
JU477754.1 | - | 966 | 100 | |||
JV597923.1 | - | 1073 | 100 | |||
JV700793.1 | - | 1038 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
