Assay Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261091.1 | NP_001248020.1 | 1248 | 112 | ||
GenBank mRNA | JU332747.1 | - | 1471 | 112 | ||
JU474166.1 | - | 1465 | 112 | |||
JV046075.1 | - | 1248 | 112 | |||
JV671754.1 | - | 1487 | 112 |
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840215.1 | XP_028696048.1 | 1487 | 112 | ||
GenBank mRNA | JV493305.1 | - | 369 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
