Assay Details
| Gene Symbol: TADA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261158.1 | NP_001248087.1 | 205 | 102 | ||
| GenBank mRNA | JU319882.1 | - | 199 | 102 | ||
| JV044064.1 | - | 205 | 102 | |||
| JV638423.1 | - | 206 | 102 | |||
More Information
Set Membership: |
|

TADA1
Genomic Map