Assay Details
| Gene Symbol: SCAMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122395.2 | XP_014977881.1 | 434 | 138 | ||
| XM_028838665.1 | XP_028694498.1 | 1717 | 138 | |||
| XM_028838666.1 | XP_028694499.1 | 1699 | 138 | |||
| XM_028838667.1 | XP_028694500.1 | 463 | 138 | |||
| XM_028838668.1 | XP_028694501.1 | 495 | 138 | |||
| GenBank mRNA | JU326213.1 | - | 286 | 138 | ||
| JU474194.1 | - | 395 | 138 | |||
| JV044148.1 | - | 391 | 138 | |||
| JV641352.1 | - | 402 | 138 | |||
| JV706141.1 | - | 396 | 138 | |||
More Information
Set Membership: |
|

SCAMP4
Genomic Map