Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028837400.1 | XP_028693233.1 | 1310 | 108 | ||
XM_028837401.1 | XP_028693234.1 | 1211 | 108 | |||
XM_028837403.1 | XP_028693236.1 | 1339 | 108 | |||
XM_028837404.1 | XP_028693237.1 | 949 | 108 | |||
XM_028837405.1 | XP_028693238.1 | 1109 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
