Assay Details
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087260.4 | XP_001087260.2 | 1328 | 137 | ||
XM_015147656.2 | XP_015003142.2 | 1196 | 137 | |||
XM_015147657.2 | XP_015003143.1 | 2851 | 137 | |||
XM_015147658.2 | XP_015003144.1 | 920 | 137 | |||
XM_015147660.2 | XP_015003146.1 | 2819 | 137 | |||
XM_015147661.2 | XP_015003147.1 | 767 | 137 | |||
XM_015147662.2 | XP_015003148.1 | 842 | 137 | |||
XM_015147663.2 | XP_015003149.1 | 789 | 137 | |||
XM_028826639.1 | XP_028682472.1 | 873 | 137 | |||
XM_028826640.1 | XP_028682473.1 | 810 | 137 | |||
XM_028826642.1 | XP_028682475.1 | 385 | 137 | |||
XM_028826643.1 | XP_028682476.1 | 381 | 137 | |||
GenBank mRNA | JU327487.1 | - | 518 | 137 | ||
JV716993.1 | - | 551 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
