Assay Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193992.2 | NP_001180921.1 | 1492 | 96 | ||
| XM_028852203.1 | XP_028708036.1 | 2270 | 96 | |||
| GenBank mRNA | JU479655.1 | - | 1118 | 96 | ||
| JV601362.1 | - | 1111 | 96 | |||
| JV635838.1 | - | 1110 | 96 | |||
More Information
Set Membership: |
|

MFSD3
Genomic Map