Assay Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193992.2 | NP_001180921.1 | 1610 | 89 | ||
| XM_015146199.2 | XP_015001685.1 | 2461 | 89 | |||
| XM_028852203.1 | XP_028708036.1 | 2388 | 89 | |||
| GenBank mRNA | JU479655.1 | - | 1236 | 89 | ||
| JV601362.1 | - | 1229 | 89 | |||
| JV635838.1 | - | 1228 | 89 | |||
More Information
Set Membership: |
|

MFSD3
Genomic Map