Assay Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109378.4 | XP_001109378.4 | 2588 | 96 | ||
XM_015136566.2 | XP_014992052.2 | 2616 | 96 | |||
XM_015136567.2 | XP_014992053.2 | 2507 | 96 | |||
XM_015136568.2 | XP_014992054.2 | 2447 | 96 | |||
XM_015136569.2 | XP_014992055.2 | 2444 | 96 | |||
XM_015136571.2 | XP_014992057.2 | 2460 | 96 | |||
XM_015136572.2 | XP_014992058.2 | 2588 | 96 | |||
XM_015136573.2 | XP_014992059.2 | 2384 | 96 | |||
XM_015136575.2 | XP_014992061.2 | 2706 | 96 | |||
XM_015136579.2 | XP_014992065.1 | 1044 | 96 | |||
XM_028847473.1 | XP_028703306.1 | 2495 | 96 | |||
XM_028847474.1 | XP_028703307.1 | 2495 | 96 | |||
XM_028847475.1 | XP_028703308.1 | 2492 | 96 | |||
XM_028847476.1 | XP_028703309.1 | 2588 | 96 | |||
XM_028847477.1 | XP_028703310.1 | 2562 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
