Assay Details
Gene Symbol: CALML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841155.1 | XP_028696988.1 | 811 | 104 | ||
XM_028841170.1 | XP_028697003.1 | 2340 | 104 | |||
GenBank mRNA | JV643896.1 | - | 146 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
