Assay Details
Gene Symbol: MLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261612.1 | NP_001248541.1 | 912 | 95 | ||
XM_015150591.2 | XP_015006077.1 | 1107 | 95 | |||
XM_015150592.2 | XP_015006078.1 | 1204 | 95 | |||
XM_028827381.1 | XP_028683214.1 | 904 | 95 | |||
XM_028827382.1 | XP_028683215.1 | 795 | 95 | |||
GenBank mRNA | JU323013.1 | - | 1036 | 95 | ||
JU323014.1 | - | 912 | 95 | |||
JV703590.1 | - | 953 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
