Assay Details
Gene Symbol: LGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194333.2 | NP_001181262.2 | 781 | 75 | ||
GenBank mRNA | JU321154.1 | - | 716 | 75 | ||
JV567313.1 | - | 774 | 75 | |||
JV602458.1 | - | 730 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
