Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151648.2 | XP_015007134.1 | 643 | 91 | ||
| XM_015151649.2 | XP_015007135.1 | 713 | 91 | |||
| XM_028829445.1 | XP_028685278.1 | 650 | 91 | |||
| XM_028829446.1 | XP_028685279.1 | 712 | 91 | |||
| GenBank mRNA | JV532973.1 | - | 203 | 91 | ||
More Information
Set Membership: |
|

RHEBL1
Genomic Map