Assay Details
| Gene Symbol: NAIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140250.2 | XP_014995736.2 | 3492 | 82 | ||
| XM_015140252.2 | XP_014995738.2 | 3484 | 82 | |||
| XM_028849494.1 | XP_028705327.1 | 3547 | 82 | |||
| XM_028849495.1 | XP_028705328.1 | 3492 | 82 | |||
More Information
Set Membership: |
|

NAIP
Genomic Map